CAS RN 39774-28-2
CAS RN 39774-28-2
IUPAC Name:
6-phenylpyridine-2-carboxylic acid
Synonyms:
6-phenylpyridine-2-carboxylic acid
Molecular Weight (g/mol):
199.21
Molecular Formula:
C12H9NO2
InChi Key:
CXOPUGLYEYDAMC-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC=CC(=N1)C1=CC=CC=C1
1
–
1
of
1
results
1
6-Phenylpyridine-2-carboxylic Acid, TRC
CAS: 39774-28-2 Molecular Formula: C12H9NO2 Molecular Weight (g/mol): 199.21 Synonym: 6-Phenylpicolinic Acid,6-Phenyl-2-pyridinecarboxylic Acid IUPAC Name: 6-phenylpyridine-2-carboxylic acid SMILES: OC(=O)c1cccc(n1)c2ccccc2