Learn More
6-Demethyl 4-Methyl Mexiletine Hydrochloride, TRC
Product Code. 30361591
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30361591 10 mg 10mg
30358885 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30361591 Supplier Toronto Research Chemicals Supplier No. TRCD29308810MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 29238-40-2
Molecular Formula C11H17NO.HCl
Molecular Weight (g/mol) 215.72
Synonym 1-(2,4-Dimethylphenoxy)-2-propanamine Hydrochloride, Mexiletine Impurity, 1-Methyl-2-(2,4-xylyloxy)-ethylamine Hydrochloride
IUPAC Name 1-(2,4-dimethylphenoxy)propan-2-amine;hydrochloride
SMILES Cl.CC(N)COc1ccc(C)cc1C
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C11H17NO.ClH/c1-8-4-5-11(9(2)6-8)13-7-10(3)12;/h4-6,10H,7,12H2,1-3H3;1H
Formula Weight 215.1077
Chemical Name or Material 6-Demethyl 4-Methyl Mexiletine Hydrochloride