CAS RN 29238-40-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
6-Demethyl 4-Methyl Mexiletine Hydrochloride, TRC
CAS: 29238-40-2 Molecular Formula: C11H17NO.HCl Molecular Weight (g/mol): 215.72 Synonym: 1-(2,4-Dimethylphenoxy)-2-propanamine Hydrochloride,Mexiletine Impurity,1-Methyl-2-(2,4-xylyloxy)-ethylamine Hydrochloride IUPAC Name: 1-(2,4-dimethylphenoxy)propan-2-amine;hydrochloride SMILES: Cl.CC(N)COc1ccc(C)cc1C