missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abcam 6-Carboxyfluorescein N-hydroxysuccinimide Ester, Fluorescent Labeling Reagent, Solid, 10 mg
Description
- Molecular Weight: 473.39 g/mol
- Form: Solid
- Applications: Fluorescent labeling of peptides, proteins and nucleotides
- Composition: 100% 6-Carboxyfluorescein-N-hydroxysuccinimide ester
- Chemical Properties: Amine-reactive succinimidyl ester carboxyfluorescein; yields hydrolysis-resistant carboxyamides
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Product Type | Protein Labeling Reagent |
| Content And Storage | Store at ambient temperature. |
| Form | Solid |
| Kit Contents | 10 mg 6-Carboxyfluorescein-N-hydroxysuccinimide ester |
| Label or Dye | 6-Carboxyfluorescein |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Target Molecule | Peptides, Proteins, Nucleotides |
| Detection Method | Fluorescence |
| Quantity | 10 mg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?