Learn More
6-APB HCl (1-Benzofuran-6-ylpropan-2-amine Hydrochloride; 6-(2-Aminopropyl)benzofuran Hydrochloride) 1.0 mg/ml in Methanol (as free base), LoGiCal
Product Code. 30457746
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457746 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30457746 Supplier LoGiCal Supplier No. LGCAMP1275.2301
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 6-APB (hydrochloride)
CAS 286834-84-2
Analyte or Component Names 6-APB (hydrochloride)
Shipping Condition Room Temperature
Molecular Formula C11 H13 N O . Cl H
Certifications/Compliance ISO 17034
Synonym 6-APB HCl, 1-Benzofuran-6-ylpropan-2-amine Hydrochloride, 6-(2-Aminopropyl)benzofuran Hydrochloride, 6-Benzofuranethanamine, α±-methyl-, hydrochloride (1:1), 6-Benzofuranethanamine, α±-methyl-, hydrochloride (9CI)
InChI Formula InChI=1S/C11H13NO.ClH/c1-8(12)6-9-2-3-10-4-5-13-11(10)7-9;/h2-5,7-8H,6,12H2,1H3;1H
Solution Type Single Solution
SMILES Cl.CC(N)Cc1ccc2ccoc2c1
IUPAC Name 1-(1-benzofuran-6-yl)propan-2-amine;hydrochloride
Solvent or Matrix Methanol
Industry Type Clinical Testing
Molecular Weight (g/mol) 211.69
Quantity 1 mL
Formula Weight 211.0764 g/mol