missing translation for 'onlineSavingsMsg'
Learn More

6-(2,3-Difluorophenyl)picolinic Acid, TRC

Product Code. 30369050
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30369050 250 mg
30379239 1 g
30377217 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30369050 Supplier Toronto Research Chemicals Supplier No. TRCD451030250MG
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 887983-10-0
Molecular Formula C12H7F2NO2
Molecular Weight (g/mol) 235.19
IUPAC Name 6-(2,3-difluorophenyl)pyridine-2-carboxylic acid
SMILES OC(=O)c1cccc(n1)c2cccc(F)c2F

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C12H7F2NO2/c13-8-4-1-3-7(11(8)14)9-5-2-6-10(15-9)12(16)17/h1-6H,(H,16,17)
Formula Weight 235.0445
Chemical Name or Material 6-(2,3-Difluorophenyl)picolinic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.