CAS RN 887983-10-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
6-(2,3-Difluorophenyl)picolinic Acid, TRC
CAS: 887983-10-0 Molecular Formula: C12H7F2NO2 Molecular Weight (g/mol): 235.19 IUPAC Name: 6-(2,3-difluorophenyl)pyridine-2-carboxylic acid SMILES: OC(=O)c1cccc(n1)c2cccc(F)c2F