missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Protein Phosphatase inhibitor 1 Antibody (SR3103), Novus Biologicals™
Shop All Bio Techne ProductsDescription
Protein Phosphatase inhibitor 1 Monoclonal antibody specifically detects Protein Phosphatase inhibitor 1 in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | Protein Phosphatase inhibitor 1 |
| Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Clone | SR3103 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:5000-1:20000, Flow Cytometry 1:20, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 |
| Formulation | PBS (pH 7.4), 150mM NaCl, 50% glycerol |
| Gene Alias | I1, I-1, IPP-1, IPP1inhibitor-1, protein phosphatase 1 regulatory subunit 1A, protein phosphatase 1, regulatory (inhibitor) subunit 1A, Protein phosphatase inhibitor 1, protein phosphatase inhibitor-1 |
| Host Species | Rabbit |
| Immunogen | A synthesized peptide derived from human Protein Phosphatase inhibitor 1 (Uniprot #: Q13522) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?