missing translation for 'onlineSavingsMsg'
Learn More

Guanoxan Sulfate, Mikromol™

Product Code. 30051724
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 30051724

Brand: Mikromol MM1880.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Guanoxan Sulfate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Guanoxan Sulfate
CAS 4-5-5714
Analyte or Component Names Guanoxan Sulfate
Shipping Condition Room Temperature
Molecular Formula 2 C10 H13 N3 O2 . H2 O4 S
Certifications/Compliance ISO 17025
Synonym Guanidine, N-[(2,3-dihydro-1,4-benzodioxin-2-yl)methyl]-, sulfate (2:1), Guanidine, (1,4-benzodioxan-2-ylmethyl)-, sulfate (2:1) (8CI), Guanidine, [(2,3-dihydro-1,4-benzodioxin-2-yl)methyl]-, sulfate (2:1) (9CI), Envacar, Guanoxan sulfate, Guanutil, NSC 108162, P 1003, P 3/01003
InChI Formula InChI=1S/2C10H13N3O2.H2O4S/c2*11-10(12)13-5-7-6-14-8-3-1-2-4-9(8)15-7;1-5(2,3)4/h2*1-4,7H,5-6H2,(H4,11,12,13);(H2,1,2,3,4)
SMILES NC(=N)NCC1COc2ccccc2O1.NC(=N)NCC3COc4ccccc4O3.OS(=O)(=O)O
IUPAC Name 1-(2,3-dihydro-1,4-benzodioxin-3-ylmethyl)guanidine;sulfuric acid
Molecular Weight (g/mol) 512.54
Quantity 50 mg
Formula Weight 512.1689 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.