missing translation for 'onlineSavingsMsg'
Learn More

Calcium (D,L)-Pantoate (Calcium (RS)-2,4-Dihydroxy-3,3-dimethylbutyrate), Mikromol™

Product Code. 30051304
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 30051304

Brand: Mikromol MM1352.05

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Calcium (D,L)-Pantoate (Calcium (RS)-2,4-Dihydroxy-3,3-dimethylbutyrate)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Calcium (D,L)-Pantoate (Calcium (RS)-2,4-Dihydroxy-3,3-dimethylbutyrate)
CAS 2769863-44-5
Analyte or Component Names Calcium (D,L)-Pantoate (Calcium (RS)-2,4-Dihydroxy-3,3-dimethylbutyrate)
Shipping Condition Room Temperature
Molecular Formula 2 C6 H11 O4 . Ca
Certifications/Compliance ISO 17025
Synonym Calcium (D,L)-Pantoate, Calcium (RS)-2,4-Dihydroxy-3,3-dimethylbutyrate
InChI Formula InChI=1S/2C6H12O4.Ca/c2*1-6(2,3-7)4(8)5(9)10;/h2*4,7-8H,3H2,1-2H3,(H,9,10);/q;;+2/p-2
SMILES [Ca+2].CC(C)(CO)C(O)C(=O)[O-].CC(C)(CO)C(O)C(=O)[O-]
IUPAC Name calcium;2,4-dihydroxy-3,3-dimethylbutanoate
Molecular Weight (g/mol) 334.38
Quantity 50 mg
Formula Weight 334.0941 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.