missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Dibenzylpiperazine.2HCl, Lipomed
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
1,4-Dibenzylpiperazine.2HCl
Specifications
Specifications
| Chemical Name or Material | 1,4-Dibenzylpiperazine Dihydrochloride |
| CAS | 2298-55-7 |
| Analyte or Component Names | 1,4-Dibenzylpiperazine Dihydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H22 N2 . 2 Cl H |
| Synonym | Piperazine, 1,4-bis(phenylmethyl)-, hydrochloride (1:2), Piperazine, 1,4-dibenzyl-, dihydrochloride (7 CI,8 CI) |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | Cl.Cl.C(N1CCN(Cc2ccccc2)CC1)c3ccccc3 |
| IUPAC Name | 1,4-dibenzylpiperazine;dihydrochloride |
| Industry Type | Pharmaceutical |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction