missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IP501 + Common metals, 500ppm: Al, Ca, Cr, Cu, Fe, K, Mg, Mn, Na, Ni, P, Pb, Si, V, Zn @ 500 μg/mL in 10% HNO3/tr. HF, VHG™
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Description
IP501 + Common metals, 500ppm: Al, Ca, Cr, Cu, Fe, K, Mg, Mn, Na, Ni, P, Pb, Si, V, Zn @ 500 μg/mL in 10% HNO3/tr. HF
Specifications
Specifications
| Chemical Name or Material | Aluminium/Calcium/Chromium/Copper/Iron/Lead/Magnesium/Manganese/Nickel/Phosphorus/Potassium/Silicon/Sodium/Vanadium/Zinc |
| CAS | 7429-90-5, 7440-70-2, 7440-47-3, 7440-50-8, 7439-89-6, 7439-92-1 |
| Methods | ICP |
| Analyte or Component Names | Aluminium/Calcium/Chromium/Copper/Iron/Lead/Magnesium/Manganese/Nickel/Phosphorus/Potassium/Silicon/Sodium/Vanadium/Zinc |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Dilute HNO3/HF |
| Quantity | 500 mL |
| Concentration or Composition (by Analyte or Components) | Aluminium 500 μg/mL/Calcium 500 μg/mL/Chromium 500 μg/mL/Copper 500 μg/mL/Iron 500 μg/mL/Lead 500 μg/mL/Magnesium 500 μg/mL/Manganese 500 μg/mL/Nickel 500 μg/mL/Phosphorus 500 μg/mL/Potassium 500 μg/mL/Silicon 500 μg/mL/Sodium 500 μg/mL/Vanadium 500 μg/mL/Zinc 500 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction