missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl-(3-iodo-1,1-dimethyl-propoxy)-dimethyL-silane, TRC

Product Code. 30179218
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179218 500 mg 500mg
30179101 100 mg 100mg
30179375 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179218 Supplier Toronto Research Chemicals Supplier No. TRCT135933500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105262-85-9
Molecular Formula C11H25OSiI
Molecular Weight (g/mol) 328.31
Synonym (1,1-Dimethylethyl)(3-iodo-1,1-dimethylpropoxy)dimethylsilane;
IUPAC Name tert-butyl-(4-iodo-2-methylbutan-2-yl)oxy-dimethylsilane
SMILES CC(C)(C)[Si](C)(C)OC(C)(C)CCI

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C11H25IOSi/c1-10(2,3)14(6,7)13-11(4,5)8-9-12/h8-9H2,1-7H3
Formula Weight 328.0719
Chemical Name or Material tert-Butyl-(3-iodo-1,1-dimethyl-propoxy)-dimethyL-silane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.