CAS RN 105262-85-9
CAS RN 105262-85-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
tert-Butyl-(3-iodo-1,1-dimethyl-propoxy)-dimethyL-silane, TRC
CAS: 105262-85-9 Molecular Formula: C11H25OSiI Molecular Weight (g/mol): 328.31 Synonym: (1,1-Dimethylethyl)(3-iodo-1,1-dimethylpropoxy)dimethylsilane; IUPAC Name: tert-butyl-(4-iodo-2-methylbutan-2-yl)oxy-dimethylsilane SMILES: CC(C)(C)[Si](C)(C)OC(C)(C)CCI