missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fmoc-tyr(2-cltrt)-oh Sodium Salt, TRC
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
Specifications
Specifications
| Chemical Name or Material | Fmoc-tyr(2-cltrt)-oh Sodium Salt |
| Analyte or Component Names | Fmoc-tyr(2-cltrt)-oh Sodium Salt |
| Shipping Condition | Room Temperature |
| Molecular Formula | C43 H33 Cl N Na O5 |
| Synonym | Sodium (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-((2-chlorophenyl)diphenylmethoxy)phenyl)propanoate |
| InChI Formula | InChI=1S/C43H34ClNO5.Na/c44-39-22-12-11-21-38(39)43(30-13-3-1-4-14-30,31-15-5-2-6-16-31)50-32-25-23-29(24-26-32)27-40(41(46)47)45-42(48)49-28-37-35-19-9-7-17-33(35)34-18-8-10-20-36(34)37;/h1-26,37,40H,27-28H2,(H,45,48)(H,46,47);/q;+1/p-1/t40-;/m0./s1 |
| Solution Type | Neat |
| SMILES | [Na]OC(=O)[C@H](Cc1ccc(OC(c2ccccc2)(c3ccccc3)c4ccccc4Cl)cc1)NC(=O)OCC5c6ccccc6c7ccccc57 |
| IUPAC Name | [(2S)-3-[4-[(2-chlorophenyl)-diphenylmethoxy]phenyl]-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoyl]oxysodium |
| Industry Type | Pharmaceutical |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction