missing translation for 'onlineSavingsMsg'
Learn More

Estradiol Benzoate, Mikromol™

Product Code. 18082510
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18082510 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18082510 Supplier Mikromol Supplier No. MM0301.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Estradiol Benzoate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Estradiol Benzoate
CAS 50-50-0
Analyte or Component Names Estradiol Benzoate
Shipping Condition Room Temperature
Molecular Formula C25 H28 O3
Certifications/Compliance ISO/IEC 17025
Synonym Estradiol Benzoate, Estradiol 3-benzoate, Agofollin, Agofollin Depot, Benovocylin, Benzhormovarine, Benzo-Gynoestryl, Benzoestrofol, Benzofoline, Benztrone, Cidirol, De Graafina, Diffollisterol, Difolliculine, Dimenformon benzoate, Diogyn B, Eston B, Estradiol benzoate, Estradiol monobenzoate, Estrogin, Femestrone, Folone, Graafina, Gynecormone, Gynformone, Hidroestron, Hormogynon, Mesalin, NSC 9566, Oestroform [BDH], Ovahormon benzoate, Ovasterol B, Ovocyclin Benzoate, Ovocyclin M, Ovocyclin MB, Pelanin benzoate, Primogyn B, Primogyn B oleosum, Progynon B, Progynon benzoate, Recthormone Oestradiol, Solestro, Unistradiol, (17beta)-Estra-1,3,5(10)-triene-3,17-diol 3-benzoate, 1,3,5(10)-Estratriene-3,17beta-diol 3-benzoate, 17beta-Estradiol 3-benzoate, 17beta-Estradiol benzoate, 3-Benzoyloxy-17beta-hydroxyestra-1,3,5(10)-triene, beta-Estradiol 3-benzoate, beta-Estradiol benzoate
InChI Formula InChI=1S/C25H28O3/c1-25-14-13-20-19-10-8-18(28-24(27)16-5-3-2-4-6-16)15-17(19)7-9-21(20)22(25)11-12-23(25)26/h2-6,8,10,15,20-23,26H,7,9,11-14H2,1H3/t20-,21-,22+,23+,25+/m1/s1
SMILES C[C@]12CC[C@H]3[C@@H](CCc4cc(OC(=O)c5ccccc5)ccc34)[C@@H]1CC[C@@H]2O
IUPAC Name [(8R,9S,13S,14S,17S)-17-hydroxy-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-3-yl] benzoate
Molecular Weight (g/mol) 376.49
Quantity 500 mg
Formula Weight 376.2038 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.