Learn More
5-tert-Butylpyrogallol, 97%, Thermo Scientific™
Product Code. 11475977
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11475977 5 g 5g
11485977 25 g 25g
11495977 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11475977 Supplier Thermo Scientific Alfa Aesar Supplier No. B25022.06
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 20481-17-8
Molecular Formula C10H14O3
Molecular Weight (g/mol) 182.219
MDL Number MFCD00059613
InChI Key HCNISNCKPIVZDX-UHFFFAOYSA-N
Synonym 5-tert-butylpyrogallol, 5-tert-butyl benzene-1,2,3-triol, 5-t-butylpyrogallol, acmc-20am1a, hcnisnckpivzdx-uhfffaoysa, 5-tert-butyl-1,2,3-benzenetriol #, 1-tert-butyl-3,4,5-trihydroxybenzene, 5-tert-butyl-1,2,3-trihydroxybenzene, 1,2,3-benzenetriol,5-1,1-dimethylethyl
PubChem CID 597592
IUPAC Name 5-tert-butylbenzene-1,2,3-triol
SMILES CC(C)(C)C1=CC(=C(C(=C1)O)O)O
Specifications
Melting Point 133°C to 136°C
Quantity 5 g
Formula Weight 182.22
Percent Purity 97%
Chemical Name or Material 5-tert-Butylpyrogallol