Learn More
5-Pyridin-4-yl-1H-indazole, TRC
Product Code. 30425924
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30425924 50 mg -
30413467 5 mg 5mg
30419512 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30425924 Supplier Toronto Research Chemicals Supplier No. TRCP99759850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 885272-55-9
Molecular Formula C12H9N3
Molecular Weight (g/mol) 195.22
IUPAC Name 5-pyridin-4-yl-2H-indazole
SMILES c1cc(ccn1)c2ccc3n[nH]cc3c2
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C12H9N3/c1-2-12-11(8-14-15-12)7-10(1)9-3-5-13-6-4-9/h1-8H,(H,14,15)
Formula Weight 195.0796
Chemical Name or Material 5-Pyridin-4-yl-1H-indazole