CAS RN 885272-55-9
CAS RN 885272-55-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5-Pyridin-4-yl-1H-indazole, TRC
CAS: 885272-55-9 Molecular Formula: C12H9N3 Molecular Weight (g/mol): 195.22 IUPAC Name: 5-pyridin-4-yl-2H-indazole SMILES: c1cc(ccn1)c2ccc3n[nH]cc3c2