missing translation for 'onlineSavingsMsg'
Learn More

5-MethylCytosine Antibody (4111), Novus Biologicals™

Product Code. 18003486 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18003486 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18003486 Supplier Novus Biologicals Supplier No. NBP242814

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Dot Blot, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunohistochemistry

5-MethylCytosine Monoclonal antibody specifically detects 5-MethylCytosine in All Species samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Dot Blot, Immunoassay, ChIP assay, Immunohistochemistry Whole-Mount.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 5-MethylCytosine
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Dot Blot, Immunoassay
Classification Monoclonal
Clone 4111
Conjugate Unconjugated
Dilution Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000, Immunohistochemistry-Frozen, Dot Blot 1:100 - 1:2000, Electron Microscopy, Methylated DNA Immunoprecipitation, Immunohistochemistry 1:50-1:500
Formulation PBS with No Preservative
Gene Alias 5-mC, 5-Methyl Cytosine, Cytosine (5-Methyl)
Host Species Mouse
Immunogen The immunogen used to generate this antibody corresponds to 5-Methylcytosine/ 5-mC
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species All species, Rat, Zebrafish
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.