missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxymethyl-2'-deoxyuridine, TRC

Product Code. 30482754
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30482754 50 mg 50mg
30474009 25 mg 25mg
30478286 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30482754 Supplier Toronto Research Chemicals Supplier No. TRCM26361050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5116-22-3
Molecular Formula C11H16N2O6
Molecular Weight (g/mol) 272.25
Synonym 5-Methoxymethyldeoxyuridine, alpha-Methoxythymidine, 2'-Deoxy-5-(methoxymethyl)uridine, 2'-Deoxy-5-(methoxymethyl)-uridine
IUPAC Name 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-(methoxymethyl)pyrimidine-2,4-dione
SMILES COCC1=CN([C@H]2C[C@H](O)[C@@H](CO)O2)C(=O)NC1=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C11H16N2O6/c1-18-5-6-3-13(11(17)12-10(6)16)9-2-7(15)8(4-14)19-9/h3,7-9,14-15H,2,4-5H2,1H3,(H,12,16,17)/t7-,8+,9+/m0/s1
Formula Weight 272.1008
Chemical Name or Material 5-Methoxymethyl-2'-deoxyuridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.