CAS RN 5116-22-3
CAS RN 5116-22-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5-Methoxymethyl-2'-deoxyuridine, TRC
CAS: 5116-22-3 Molecular Formula: C11H16N2O6 Molecular Weight (g/mol): 272.25 Synonym: 5-Methoxymethyldeoxyuridine,alpha-Methoxythymidine,2'-Deoxy-5-(methoxymethyl)uridine,2'-Deoxy-5-(methoxymethyl)-uridine IUPAC Name: 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-(methoxymethyl)pyrimidine-2,4-dione SMILES: COCC1=CN([C@H]2C[C@H](O)[C@@H](CO)O2)C(=O)NC1=O