Learn More
5-Methoxy-2-methylbenzothiazole, 97%, Thermo Scientific Chemicals
Product Code. 11405167
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11405167 5 g 5g
11495157 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11405167 Supplier Thermo Scientific Alfa Aesar Supplier No. B22584.06
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2941-69-7
Molecular Formula C9H9NOS
Molecular Weight (g/mol) 179.24
MDL Number MFCD00082664
InChI Key SAQMNBWVOKYKPZ-UHFFFAOYSA-N
PubChem CID 76252
IUPAC Name 5-methoxy-2-methyl-1,3-benzothiazole
SMILES COC1=CC=C2SC(C)=NC2=C1
Specifications
Melting Point 34°C to 38°C
Refractive Index 1.61
Quantity 5 g
Beilstein 124232
Synonym 5-methoxy-2-methylbenzothiazole, 5-methoxy-2-methylbenzo d thiazole, benzothiazole, 5-methoxy-2-methyl, 2-methyl-5-methoxybenzothiazole, 2-methyl-5-methoxy-benzothiazole, acmc-209h8o, cambridge id 5150005, benzothiazole,5-methoxy-2-methyl, 2-methyl-5-methoxy-1,3-benzothiazole, 2-methyl-5-methoxy-1, 3-benzothiazole
Formula Weight 179.24
Percent Purity 97%
Chemical Name or Material 5-Methoxy-2-methylbenzothiazole