accessibility menu, dialog, popup

UserName

CAS RN 2941-69-7

CH3OSH3CN

CAS RN 2941-69-7

IUPAC Name: 5-methoxy-2-methyl-1,3-benzothiazole
Synonyms: 5-methoxy-2-methyl-1,3-benzothiazole
Molecular Weight (g/mol): 179.24
Molecular Formula: C9H9NOS
InChi Key: SAQMNBWVOKYKPZ-UHFFFAOYSA-N
SMILES: COC1=CC=C2SC(C)=NC2=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)