Learn More
5-hydroxy Chlorosulfuron, TRC
Product Code. 30180234
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180234 1 mg 1mg
30180259 5 mg 5mg
30179834 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30180234 Supplier Toronto Research Chemicals Supplier No. TRCH8394051MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 81123-38-8
Molecular Formula C12 H12 Cl N5 O5 S
Molecular Weight (g/mol) 373.77
Synonym 2-Chloro-5-hydroxy-N-[[(4-methoxy-6-methyl-1,3,5-triazin-2-yl)amino]carbonyl]benzenesulfonamide, N 5754
IUPAC Name 1-(2-chloro-5-hydroxyphenyl)sulfonyl-3-(4-methoxy-6-methyl-1,3,5-triazin-2-yl)urea
SMILES COc1nc(C)nc(NC(=O)NS(=O)(=O)c2cc(O)ccc2Cl)n1
Specifications
Quantity 1 mg
InChI Formula InChI=1S/C12H12ClN5O5S/c1-6-14-10(17-12(15-6)23-2)16-11(20)18-24(21,22)9-5-7(19)3-4-8(9)13/h3-5,19H,1-2H3,(H2,14,15,16,17,18,20)
Formula Weight 373.0248
Chemical Name or Material Chlorsulfuron-5-hydroxy