CAS RN 81123-38-8
CAS RN 81123-38-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5-hydroxy Chlorosulfuron, TRC
CAS: 81123-38-8 Molecular Formula: C12 H12 Cl N5 O5 S Molecular Weight (g/mol): 373.77 Synonym: 2-Chloro-5-hydroxy-N-[[(4-methoxy-6-methyl-1,3,5-triazin-2-yl)amino]carbonyl]benzenesulfonamide,N 5754 IUPAC Name: 1-(2-chloro-5-hydroxyphenyl)sulfonyl-3-(4-methoxy-6-methyl-1,3,5-triazin-2-yl)urea SMILES: COc1nc(C)nc(NC(=O)NS(=O)(=O)c2cc(O)ccc2Cl)n1