missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-HT2B Antibody (CL14576) [CoraFluor™ 1], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-26870CL1
This item is not returnable.
View return policy
Description
5-HT2B Monoclonal antibody specifically detects 5-HT2B in Human samples. It is validated for Western Blot
Specifications
| 5-HT2B | |
| Monoclonal | |
| CoraFluor 1 | |
| 5-HT(2B), 5-HT-2B, 5-HT2B5-HT 2B receptor, 5-hydroxytryptamine (serotonin) receptor 2B, 5-hydroxytryptamine 2B receptor, 5-hydroxytryptamine receptor 2B, Serotonin receptor 2B | |
| This antibody was developed using a recombinant protein derived from P41595, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 3357 | |
| Store at 4⪚C in the dark. Do not freeze. | |
| IgG1 |
| Western Blot | |
| CL14576 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction