missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[5-[(Dimethyl-amino)methyl]furan-2-yl]methanol Hydrochloride, Mikromol™
Description
[5-[(Dimethyl-amino)methyl]furan-2-yl]methanol Hydrochloride
Specifications
Specifications
| Chemical Name or Material | (5-[(dimethylamino)methyl]furan-2-yl)methanol Hydrochloride |
| CAS | 81074-81-9 |
| Analyte or Component Names | (5-[(dimethylamino)methyl]furan-2-yl)methanol Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C8 H13 N O2 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Ranitidine Hydrochloride Imp. F (EP), Ranitidine Imp. F (EP), [5-[(Dimethyl-amino)methyl]furan-2-yl]methanol Hydrochloride, Ranitidine Hydrochloride Imp. F (EP) as Hydrochloride, Ranitidine Hydrochloride Impurity F as Hydrochloride, Ranitidine Impurity F as Hydrochloride |
| InChI Formula | InChI=1S/C8H13NO2.ClH/c1-9(2)5-7-3-4-8(6-10)11-7;/h3-4,10H,5-6H2,1-2H3;1H |
| SMILES | Cl.CN(C)Cc1oc(CO)cc1 |
| IUPAC Name | [5-[(dimethylamino)methyl]furan-2-yl]methanol;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?