missing translation for 'onlineSavingsMsg'
Learn More

5-cis-Tetradecenoyl Carnitine, TRC

Product Code. 30427920
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
10 mg
Unit Size:
10mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30427920 1 mg -
30426796 2.5 mg 2.5mg
30419543 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30427920 Supplier Toronto Research Chemicals Supplier No. TRCT2914251MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1186416-86-3
Molecular Formula C21 H39 N O4
Molecular Weight (g/mol) 369.54
Synonym 1-Propanaminium, 3-carboxy-N,N,N-trimethyl-2-[[(5Z)-1-oxo-5-tetradecen-1-yl]oxy]-, inner salt, (2R)-, 5-cis-Tetradecenoyl L-carnitine, 5-cis-Tetradecenoyl carnitine, (2R)-3-Carboxy-N,N,N-trimethyl-2-[[(5Z)-1-oxo-5-tetradecen-1-yl]oxy]-1-propanaminium inner salt
IUPAC Name (3R)-3-[(Z)-tetradec-5-enoyl]oxy-4-(trimethylazaniumyl)butanoate
SMILES CCCCCCCC\C=C/CCCC(=O)O[C@H](CC(=O)[O-])C[N+](C)(C)C

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C21H39NO4/c1-5-6-7-8-9-10-11-12-13-14-15-16-21(25)26-19(17-20(23)24)18-22(2,3)4/h12-13,19H,5-11,14-18H2,1-4H3/b13-12-/t19-/m1/s1
Formula Weight 369.2879
Percent Purity >95%
Chemical Name or Material 5-cis-Tetradecenoyl L-Carnitine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.