CAS RN 1186416-86-3
CAS RN 1186416-86-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5-cis-Tetradecenoyl Carnitine, TRC
CAS: 1186416-86-3 Molecular Formula: C21 H39 N O4 Molecular Weight (g/mol): 369.54 Synonym: 1-Propanaminium, 3-carboxy-N,N,N-trimethyl-2-[[(5Z)-1-oxo-5-tetradecen-1-yl]oxy]-, inner salt, (2R)-,5-cis-Tetradecenoyl L-carnitine,5-cis-Tetradecenoyl carnitine,(2R)-3-Carboxy-N,N,N-trimethyl-2-[[(5Z)-1-oxo-5-tetradecen-1-yl]oxy]-1-propanaminium inner salt IUPAC Name: (3R)-3-[(Z)-tetradec-5-enoyl]oxy-4-(trimethylazaniumyl)butanoate SMILES: CCCCCCCC\C=C/CCCC(=O)O[C@H](CC(=O)[O-])C[N+](C)(C)C