missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-6-hydroxynicotinic acid, 98%

Product Code. 11482378
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11482378

Brand: Thermo Scientific Alfa Aesar A15737.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 54127-63-8 | C6H3ClNO3 | 172.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54127-63-8
Molecular Formula C6H3ClNO3
Molecular Weight (g/mol) 172.54
MDL Number MFCD00052950
InChI Key OLTRUTPHSBQWAZ-UHFFFAOYSA-M
Synonym 5-chloro-6-hydroxynicotinic acid, 3-chloro-2-hydroxypyridine-5-carboxylic acid, 5-chloro-6-hydroxypyridine-3-carboxylic acid, 5-chloro-6-hydroxy-3-pyridinecarboxylic acid, 5-chloro-6-hydroxynicotinicacid, 3-pyridinecarboxylic acid, 5-chloro-1,6-dihydro-6-oxo, 5-chloro-6-oxo-1,6-dihydropyridine-3-carboxylic acid, pubchem10618, maybridge1_008981, acmc-1b1yw
PubChem CID 599541
IUPAC Name 5-chloro-6-oxo-1H-pyridine-3-carboxylic acid
SMILES [O-]C(=O)C1=CNC(=O)C(Cl)=C1

Specifications

Melting Point >300°C
Quantity 25 g
Formula Weight 173.56
Percent Purity 98%
Chemical Name or Material 5-Chloro-6-hydroxynicotinic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.