CAS RN 54127-63-8
CAS RN 54127-63-8
IUPAC Name:
5-chloro-6-oxo-1,6-dihydropyridine-3-carboxylate
Synonyms:
5-chloro-6-oxo-1H-pyridine-3-carboxylate
Molecular Weight (g/mol):
172.54
Molecular Formula:
C6H3ClNO3
InChi Key:
OLTRUTPHSBQWAZ-UHFFFAOYSA-M
SMILES:
[O-]C(=O)C1=CNC(=O)C(Cl)=C1
1
–
1
of
1
results
1
5-Chloro-6-hydroxynicotinic acid, 98%
CAS: 54127-63-8 Molecular Formula: C6H3ClNO3 Molecular Weight (g/mol): 172.54 MDL Number: MFCD00052950 InChI Key: OLTRUTPHSBQWAZ-UHFFFAOYSA-M Synonym: 5-chloro-6-hydroxynicotinic acid,3-chloro-2-hydroxypyridine-5-carboxylic acid,5-chloro-6-hydroxypyridine-3-carboxylic acid,5-chloro-6-hydroxy-3-pyridinecarboxylic acid,5-chloro-6-hydroxynicotinicacid,3-pyridinecarboxylic acid, 5-chloro-1,6-dihydro-6-oxo,5-chloro-6-oxo-1,6-dihydropyridine-3-carboxylic acid,pubchem10618,maybridge1_008981,acmc-1b1yw PubChem CID: 599541 IUPAC Name: 5-chloro-6-oxo-1H-pyridine-3-carboxylic acid SMILES: [O-]C(=O)C1=CNC(=O)C(Cl)=C1