missing translation for 'onlineSavingsMsg'
Få mere at vide

5-Chloro-3-methyl-1-phenyl-1H-pyrazole-4-carboxaldehyde, 97%, Thermo Scientific™

Artikelnummer. 15483826
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
1 g
5 g
Pakningsstørrelse:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
15483826 1 g 1g
15493826 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15483826 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. H57939.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
Se alternative produkter

Denne vare kan ikke returneres. Se returpolicy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 947-95-5
Molecular Formula C11H9ClN2O
Molecular Weight (g/mol) 220.656
MDL Number MFCD00100755
InChI Key DKZPJLZXLKAMDO-UHFFFAOYSA-N
Synonym 5-chloro-3-methyl-1-phenyl-1h-pyrazole-4-carbaldehyde, 5-chloro-3-methyl-1-phenyl pyrazole-4-alldehyde, 5-chloro-4-formyl-3-methyl-1-phenylpyrazole, acmc-20agcp, 5-chloro-3-methyl-1-phenyl-4-pyrazolecarboxaldehyde, 5-chloro-3-methyl-1-phenyl pyrazole-4-aldehyde, chloro-3-methyl-1-phenylpyrazole-4-carboxaldehyde, 5-chloro-3-methyl-1-phenylpyrazole-4-carboxaldehyde, 2-phenyl-3-chloro-5-methyl-2h-pyrazole-4-carbaldehyde, pyrazole-4-carboxaldehyde, 5-chloro-3-methyl-1-phenyl
PubChem CID 613027
IUPAC Name 5-chloro-3-methyl-1-phenylpyrazole-4-carbaldehyde
SMILES CC1=NN(C(=C1C=O)Cl)C2=CC=CC=C2

Tekniske data

Melting Point 145°C to 148°C
Quantity 1 g
Formula Weight 220.65
Percent Purity 97%
Chemical Name or Material 5-Chloro-3-methyl-1-phenyl-1H-pyrazole-4-carboxaldehyde
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.