accessibility menu, dialog, popup

UserName

CH3NNClO

CAS RN 947-95-5

IUPAC Name: 5-chloro-3-methyl-1-phenyl-1H-pyrazole-4-carbaldehyde
Synonyms: 5-chloro-3-methyl-1-phenylpyrazole-4-carbaldehyde
Molecular Weight (g/mol): 220.66
Molecular Formula: C11H9ClN2O
InChi Key: DKZPJLZXLKAMDO-UHFFFAOYSA-N
SMILES: CC1=NN(C(Cl)=C1C=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results