missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-(methylamino)benzophenone, TRC

Product Code. 30012810
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012810 1 g 1g
30006652 100 g 100g
30010162 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012810 Supplier Toronto Research Chemicals Supplier No. TRCC3690201G
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1022-13-5
Molecular Formula C14 H12 Cl N O
Molecular Weight (g/mol) 245.7
Synonym [5-Chloro-2-(methylamino)phenyl]phenylmethanone, Temazepam Imp. A (EP), Diazepam Imp. D (EP), 5-Chloro-2-(methylamino)benzophenone
IUPAC Name [5-chloro-2-(methylamino)phenyl]-phenylmethanone
SMILES CNc1ccc(Cl)cc1C(=O)c2ccccc2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C14H12ClNO/c1-16-13-8-7-11(15)9-12(13)14(17)10-5-3-2-4-6-10/h2-9,16H,1H3
Formula Weight 245.0607
Percent Purity >95%
Chemical Name or Material 5-Chloro-2-Methylaminobenzophenone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.