missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-7-methyl-1H-indazole, 97%, Thermo Scientific Chemicals

Product Code. 11316819
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11316819

Brand: Thermo Scientific Alfa Aesar H26753.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 156454-43-2
Molecular Formula C8H7BrN2
Molecular Weight (g/mol) 211.062
MDL Number MFCD03990484
InChI Key OUNQFZFHLJLFAR-UHFFFAOYSA-N
PubChem CID 1382033
IUPAC Name 5-bromo-7-methyl-1H-indazole
SMILES CC1=C2C(=CC(=C1)Br)C=NN2

Specifications

Melting Point 183°C to 186°C
Quantity 1 g
Synonym 1h-indazole, 5-bromo-7-methyl, indazole, 5-bromo-7-methyl, pubchem20590, acmc-209zze, 5-bromo-7-methylindazole, ksc495o1l, 2h-indazole,5-bromo-7-methyl, 1h-indazole,5-bromo-7-methyl, 2h-indazole, 5-bromo-7-methyl, en
Formula Weight 211.06
Percent Purity 97%
Chemical Name or Material 5-Bromo-7-methyl-1H-indazole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.