missing translation for 'onlineSavingsMsg'
Learn More

5-Acetyl-1,3-phenylene Bis(dimethylcarbamate), Mikromol™

Product Code. 30052047
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30052047 100 mg 100mg
30050878 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30052047 Supplier Mikromol Supplier No. MM0773.05

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

5-Acetyl-1,3-phenylene Bis(dimethylcarbamate)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Acetyl-1,3-phenylene Bis(dimethylcarbamate)
CAS 81732-48-1
Analyte or Component Names 5-Acetyl-1,3-phenylene Bis(dimethylcarbamate)
Shipping Condition Room Temperature
Molecular Formula C14 H18 N2 O5
Certifications/Compliance ISO 17025
Synonym Bambuterol Hydrochloride Imp. E (EP), Bambuterol Imp. E (EP), 5-Acetyl-1,3-phenylene Bis(dimethylcarbamate), Bambuterol Hydrochloride Impurity E, Bambuterol Impurity E
InChI Formula InChI=1S/C14H18N2O5/c1-9(17)10-6-11(20-13(18)15(2)3)8-12(7-10)21-14(19)16(4)5/h6-8H,1-5H3
SMILES CN(C)C(=O)Oc1cc(OC(=O)N(C)C)cc(c1)C(=O)C
IUPAC Name [3-acetyl-5-(dimethylcarbamoyloxy)phenyl] N,N-dimethylcarbamate
Molecular Weight (g/mol) 294.3
Quantity 100 mg
Formula Weight 294.1216 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.