missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-[(2RS)-2-[[2-(2-Ethoxyphenoxy)ethyl]amino]propyl]-2-methoxybenzenesulfonamide Hydrochloride (Tamsulosin Racemate Hydrochloride), Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
5-[(2RS)-2-[[2-(2-Ethoxyphenoxy)ethyl]amino]propyl]-2-methoxybenzenesulfonamide Hydrochloride (Tamsulosin Racemate Hydrochloride)
Specifications
Specifications
| Chemical Name or Material | 5-[(2RS)-2-[(2-(2-Ethoxyphenoxy)ethyl)amino]propyl]-2-methoxybenzenesulfonamide Hydrochloride (Tamsulosin Racemate Hydrochloride) |
| CAS | 80223-99-0 |
| Analyte or Component Names | 5-[(2RS)-2-[(2-(2-Ethoxyphenoxy)ethyl)amino]propyl]-2-methoxybenzenesulfonamide Hydrochloride (Tamsulosin Racemate Hydrochloride) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H28 N2 O5 S . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Tamsulosin Hydrochloride Imp. G (EP), Tamsulosin Imp. G (EP), Racemic Tamsulosin Hydrochloride, 5-[(2RS)-2-[[2-(2-Ethoxyphenoxy)ethyl]amino]propyl]-2-methoxybenzenesulfonamide Hydrochloride, Tamsulosin Hydrochloride Imp. G (EP) as Racemate Hydrochloride, Tamsulosin Racemate Hydrochloride, Benzenesulfonamide, 5-[2-[[2-(2-ethoxyphenoxy)ethyl]amino]propyl]-2-methoxy-, hydrochloride (1:1), Benzenesulfonamide, 5-[2-[[2-(2-ethoxyphenoxy)ethyl]amino]propyl]-2-methoxy-, monohydrochloride (9CI), LY 253352, YM 12617, dl-Tamsulosin hydrochloride, 5-[(2RS)-2-[[2-(2-Ethoxyphenoxy)ethyl]amino]propyl]-2-methoxybenzenesulfonamide Hydrochloride (Tamsulosin Racemate Hydrochloride), Tamsulosin Hydrochloride Impurity G as Racemate Hydrochloride, Tamsulosin Impurity G as Racemate Hydrochloride |
| InChI Formula | InChI=1S/C20H28N2O5S.ClH/c1-4-26-17-7-5-6-8-18(17)27-12-11-22-15(2)13-16-9-10-19(25-3)20(14-16)28(21,23)24;/h5-10,14-15,22H,4,11-13H2,1-3H3,(H2,21,23,24);1H |
| SMILES | Cl.CCOc1ccccc1OCCNC(C)Cc2ccc(OC)c(c2)S(=O)(=O)N |
| IUPAC Name | 5-[2-[2-(2-ethoxyphenoxy)ethylamino]propyl]-2-methoxybenzenesulfonamide;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction