missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(4S)-2-[Carboxy[(phenoxy-acetyl)amino]methyl]-5,5-dimethylthiazolidine-4-carboxylic Acid (Penicilloic Acids of Phenoxymethylpenicillin), Mikromol™
Description
(4S)-2-[Carboxy[(phenoxy-acetyl)amino]methyl]-5,5-dimethylthiazolidine-4-carboxylic Acid (Penicilloic Acids of Phenoxymethylpenicillin)
Specifications
Specifications
| Chemical Name or Material | (4S)-2-[Carboxy[(phenoxy-acetyl)amino]methyl]-5,5-dimethylthiazolidine-4-carboxylic Acid (Penicilloic Acids of Phenoxymethylpenicillin) |
| CAS | 1049-84-9 |
| Analyte or Component Names | (4S)-2-[Carboxy[(phenoxy-acetyl)amino]methyl]-5,5-dimethylthiazolidine-4-carboxylic Acid (Penicilloic Acids of Phenoxymethylpenicillin) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H20 N2 O6 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Phenoxymethylpenicillin Potassium Imp. E (EP), Phenoxymethylpenicillin Imp. E (EP), (4S)-2-[Carboxy[(phenoxy-acetyl)amino]methyl]-5,5-dimethylthiazolidine-4-carboxylic Acid, Penicilloic Acids of Phenoxymethylpenicillin, Phenoxymethylpenicillin Potassium Impurity E, Phenoxymethylpenicillin Impurity E |
| InChI Formula | InChI=1S/C16H20N2O6S/c1-16(2)12(15(22)23)18-13(25-16)11(14(20)21)17-10(19)8-24-9-6-4-3-5-7-9/h3-7,11-13,18H,8H2,1-2H3,(H,17,19)(H,20,21)(H,22,23)/t11?,12-,13?/m0/s1 |
| SMILES | CC1(C)SC(N[C@H]1C(=O)O)C(NC(=O)COc2ccccc2)C(=O)O |
| IUPAC Name | (4S)-2-[carboxy-[(2-phenoxyacetyl)amino]methyl]-5,5-dimethyl-1,3-thiazolidine-4-carboxylic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?