Learn More
(4R)-4-(Isopropylamino)-2-(3-methoxypropyl)-3,4-dihydro-2H-thieno[3,2-e]-1,2-thiazine-6-sulfonamide 1,1-Dioxide, Mikromol™
Description
(4R)-4-(Isopropylamino)-2-(3-methoxypropyl)-3,4-dihydro-2H-thieno[3,2-e]-1,2-thiazine-6-sulfonamide 1,1-Dioxide, Mikromol
Specifications
Specifications
| Chemical Name or Material | (4R)-4-(Isopropylamino)-2-(3-methoxypropyl)-3,4-dihydro-2H-thieno[3,2-e]-1,2-thiazine-6-sulfonamide-1,1-dioxide |
| CAS | 2985481-46-5 |
| Analyte or Component Names | (4R)-4-(Isopropylamino)-2-(3-methoxypropyl)-3,4-dihydro-2H-thieno[3,2-e]-1,2-thiazine-6-sulfonamide-1,1-dioxide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H23 N3 O5 S3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | (4R)-4-(Isopropylamino)-2-(3-methoxypropyl)-1,1-dioxo-3,4-dihydrothieno[3,2-e]thiazine-6-sulfonamide, (4R)-4-(Isopropylamino)-2-(3-methoxypropyl)-3,4-dihydro-2H-thieno[3,2-e]-1,2-thiazine-6-sulfonamide-1,1-dioxide, N-Desethyl N-Isopropyl Brinzolamide |
| InChI Formula | InChI=1S/C13H23N3O5S3/c1-9(2)15-11-8-16(5-4-6-21-3)24(19,20)13-10(11)7-12(22-13)23(14,17)18/h7,9,11,15H,4-6,8H2,1-3H3,(H2,14,17,18)/t11-/m0/s1 |
| Solution Type | Neat |
| SMILES | COCCCN1C[C@H](NC(C)C)c2cc(sc2S1(=O)=O)S(=O)(=O)N |
| Show More |