missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Michigan Residual Solvents Mixture Kit 703 Inhaled Products 1-2100 μg/mL in Triacetin, Dr. Ehrenstorfer
Click to view available options
Quantity:
4 x 1 mL
Unit Size:
4mL
Specifications
Specifications
| CAS | 107-06-2, 67-64-1, 75-05-8, 71-43-2, 67-66-3, 75-09-2 |
| Quantity | 4 x 1 mL |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Triacetin |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction