missing translation for 'onlineSavingsMsg'
Learn More

4H-Thieno[3,2-b]pyrrole-5-carboxylic acid, 97%, Thermo Scientific™

Product Code. p-3110655
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10249612

Brand: Thermo Scientific Maybridge CC39001CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 39793-31-2
Molecular Formula C7H5NO2S
Molecular Weight (g/mol) 167.182
MDL Number MFCD07368542
InChI Key PMHDSACGRKBACK-UHFFFAOYSA-N
Synonym 4h-thieno 3,2-b pyrrole-5-carboxylic acid, 4h-thieno 3,2-b pyrole-5-carboxylic acid, 4wl, d-amino acid oxidase inhibitor, 4h-thieno 3,2-b pyrrole-5-carboxylicacid, thiopheno 3,2-b pyrrole-5-carboxylic acid, 4h-thieno 2,3-d pyrrole-5-carboxylic acid, 4h-thieno 3 pound not2-b pyrrole-5-carboxylic acid
PubChem CID 4961254
IUPAC Name 4H-thieno[3,2-b]pyrrole-5-carboxylic acid
SMILES C1=CSC2=C1NC(=C2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 167.19
Percent Purity 97%
Chemical Name or Material 4H-Thieno[3, 2-b]pyrrole-5-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.