accessibility menu, dialog, popup

UserName

SNHOOH

CAS RN 39793-31-2

IUPAC Name: 4H-thieno[3,2-b]pyrrole-5-carboxylic acid
Synonyms: 4H-thieno[3,2-b]pyrrole-5-carboxylic acid
Molecular Weight (g/mol): 167.18
Molecular Formula: C7H5NO2S
InChi Key: PMHDSACGRKBACK-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC2=C(N1)C=CS2
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results