missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Assay type: Cell-based
- Detection method: Flow cytometry-fluorescent
- Applications: Flow Cyt
- Sample types: Suspension cells, Adherent cells
- Reactive species: Mammals
- Results type: Quantitative
- Key features: Labeling in < 4 hrs with 30 secs hands-on time; 100% antibody recovery
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Content And Storage | Store at -20°C. |
| For Use With (Equipment) | Flow Cytometer |
| Format | Kit |
| No. of Reactions | 500 Tests |
| Reactivity | Mammals |
| Shipping Condition | Ambient |
| Target Molecule | Extracellular Vesicles |
| Detection Method | Fluorescence |
| Cross Reactivity | Mammals |
| Sample Type | Suspension cells, Adherent cells |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?