Learn More
4-(Trifluoromethyl)benzamide, 98+%, Thermo Scientific™
Product Code. 11420713
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11420713 25 g 25g
11410713 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11420713 Supplier Thermo Scientific Alfa Aesar Supplier No. A12746.14
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1891-90-3
Molecular Formula C8H6F3NO
Molecular Weight (g/mol) 189.14
MDL Number MFCD00007998
InChI Key WEJHBEDHLLBJFW-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzamide, p-trifluoromethylbenzamide, p-trifluoromethyl benzamide, benzamide, 4-trifluoromethyl, pubchem5482, acmc-20a3xk, 4-carbamoylbenzotrifluoride, 4-trifluoromethyl-benzamide, 4-cf3-c6h4conh2, n-4-trifluoromethyl benzamide
PubChem CID 74684
IUPAC Name 4-(trifluoromethyl)benzamide
SMILES NC(=O)C1=CC=C(C=C1)C(F)(F)F
Spezifikation
Melting Point 182°C to 186°C
Odor Odorless
Quantity 25 g
Beilstein 2098929
Formula Weight 189.14
Percent Purity ≥98%
Chemical Name or Material 4-(Trifluoromethyl)benzamide