missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Pyridoxic Acid, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
4-Pyridoxic Acid
Specifications
Specifications
| Chemical Name or Material | 4-Pyridoxic Acid |
| CAS | 82-82-6 |
| Analyte or Component Names | 4-Pyridoxic Acid |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C8 H9 N O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 4-Pyridoxic Acid |
| InChI Formula | InChI=1S/C8H9NO4/c1-4-7(11)6(8(12)13)5(3-10)2-9-4/h2,10-11H,3H2,1H3,(H,12,13) |
| SMILES | Cc1ncc(CO)c(C(=O)O)c1O |
| IUPAC Name | 3-hydroxy-5-(hydroxymethyl)-2-methylpyridine-4-carboxylic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction