missing translation for 'onlineSavingsMsg'
Learn More

Abcam 4-Oxo-2-nonenal, TRPA1 Activator, >98% Purity, Liquid, 5 mg

Product Code. 32173564
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32173564 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32173564 Supplier Abcam Supplier No. ab1208785mg
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent lipid peroxidation product and TRPA1 activator with >98% purity.

  • Molecular weight: 154.21 g/mol
  • Purity: >98%
  • Form: Liquid
  • Storage buffer: Constituents: 99.5% Methyl acetate, 0.5% (E)-4-oxonon-2-enal
  • CAS number: 103560-62-9
  • Activity: Lipid peroxidation product that modifies histidine and lysine residues on proteins, causes protein cross-linking, and acts as a TRPA1 activator (EC50 = 1.9 μM)
  • Stability: Active in vivo
  • Storage conditions: Store at -80°C
  • Shipping conditions: Dry Ice

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Content And Storage Store at -80°C.
Format Vial
Form Liquid
Shipping Condition Dry Ice
Substrate Properties Chemical Substrate
Target Molecule TRPA1
Formulation 99.5% Methyl acetate, 0.5% (E)-4-oxonon-2-enal
Quantity 5 mg
For Use With (Application) TRPA1 activation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.