missing translation for 'onlineSavingsMsg'
Learn More

4-Octylphenol Monoethoxylate, TRC

Product Code. 30008876
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008876 1 g 1g
30008390 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30008876 Supplier Toronto Research Chemicals Supplier No. TRCO2937701G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51437-89-9
Molecular Formula C16 H26 O2
Molecular Weight (g/mol) 250.38
Synonym 2-(4-Octylphenoxy)ethanol, 2-(p-Octylphenoxy)ethanol, Ethylene Glycol p-Octylphenyl Ether;
IUPAC Name 2-(4-octylphenoxy)ethanol
SMILES CCCCCCCCc1ccc(OCCO)cc1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C16H26O2/c1-2-3-4-5-6-7-8-15-9-11-16(12-10-15)18-14-13-17/h9-12,17H,2-8,13-14H2,1H3
Formula Weight 250.1933
Percent Purity >95%
Chemical Name or Material 4-Octylphenol-mono-ethoxylate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.