CAS RN 51437-89-9
CAS RN 51437-89-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Octylphenol Monoethoxylate, TRC
CAS: 51437-89-9 Molecular Formula: C16 H26 O2 Molecular Weight (g/mol): 250.38 Synonym: 2-(4-Octylphenoxy)ethanol,2-(p-Octylphenoxy)ethanol,Ethylene Glycol p-Octylphenyl Ether; IUPAC Name: 2-(4-octylphenoxy)ethanol SMILES: CCCCCCCCc1ccc(OCCO)cc1