missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenol Sodium Salt Dihydrate, TRC

Product Code. 30011525
Change view
Click to view available options
Quantity:
1 g
2.5 g
Unit Size:
1g
2.50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011525 1 g 1g
30138763 2.5 g 2.50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011525 Supplier Toronto Research Chemicals Supplier No. TRCN5044481G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66924-59-2
Formula Weight 197.03
InChI Formula InChI=1S/C6H5NO3.Na.2H2O/c8-6-3-1-5(2-4-6)7(9)10;;;/h1-4,8H;;2*1H2/q;+1;;/p-1
IUPAC Name sodium;4-nitrophenolate;dihydrate
Molecular Formula C6H4NNaO3 . 2(H2O)
Molecular Weight (g/mol) 161.09
SMILES O.O.[Na+].[O-]c1ccc(cc1)[N+](=O)[O-]
Synonym Sodium p-Nitrophenolate Dihydrate, 4-Nitro-phenol Sodium Salt Dihydrate
Chemical Name or Material 4-Nitrophenol Sodium Salt Dihydrate

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.